Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brij 35, 30% (w/w) Aqueous Wetting Agent, Ricca Chemical
SDP

Catalog No. 114532 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1145-32 1 L
114516 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 1145-32 Supplier Ricca Chemical Company Supplier No. 114532
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material Brij 35
Color Colorless
Name Note Wetting Agent
CAS 7732-18-5, 9002-92-0
Physical Form Liquid
CAS Min % 69.0, 28.99
CAS Max % 72.0, 30.17
Concentration 30% (w/w)
Quantity 1 L
Packaging Natural Poly Bottle
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.