Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Brilliant Blue R, pure

Catalog No. AC191491000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC191491000 100 g Glass bottle
AC191490050 5 g Glass bottle
AC191490250 25 g Glass bottle
AC191495000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC191491000 Supplier Thermo Scientific Chemicals Supplier No. 191491000
Add to Cart

CAS: 6104-59-2 | C45H44N3NaO7S2 | 825.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6104-59-2
Molecular Formula C45H44N3NaO7S2
Molecular Weight (g/mol) 825.97
MDL Number MFCD00041762
InChI Key HAEGGLCXLFWTBE-UHFFFAOYSA-M
Synonym Acid Blue 83, C.I. 42660, Coomassie™ Brilliant Blue R-250
PubChem CID 61365
IUPAC Name sodium;3-[[4-[[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate
SMILES [Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1

Specifications

Absorbance 700 (at lambda min.)
Melting Point 174.0°C to 180.0°C
Absorption Ratio 1 to 1.2
Solubility Information Solubility in water: slightly soluble. Other solubilities: practically insoluble in ethanol
Infrared Spectrum Authentic
Lambda Max 588 to 596nm (in 50:50 ethanol:water)
Formula Weight 825.98
Grade Pure
Packaging Glass bottle
Color Bordeaux to Brown/Purple
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material Brilliant Blue R, Pure
Show More Show Less
EINECSNumber 228-060-5
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.