Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brilliant Green, MP Biomedicals™

Catalog No. MP215051625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215051625 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215051625 Supplier MP Biomedicals, Inc Supplier No. 0215051625
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Also called emerald green or malachite green. Brilliant Green, MP Biomedicals is one of the triarylmethane dyes of the malachite-green series. It occurs as small, shiny, golden crystals soluble in water or alcohol.

Brilliant green is used in dilute solution as a topical antiseptic. It is effective against gram-positive microorganisms. It has also been used to dye silk and wool.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 633-03-4
Molecular Formula C27H34N2O4S
Molecular Weight (g/mol) 482.639
InChI Key NNBFNNNWANBMTI-UHFFFAOYSA-M
Synonym Astradiamant green GX, Basic Green 1, Diamond Green, Emerald Green, Ethyl Green, Malachite Green G, Solid Green JO
PubChem CID 12449
IUPAC Name [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate
SMILES CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]

Specifications

Solubility Information Soluble in water (1mg/mL), ethanol, methanol.
Quantity 25 g
Physical Form Metallic Crystalline Powder
Formula Weight 482.65
Grade Certified Biological Stain
Chemical Name or Material Brilliant Green
Recommended Storage Room Temperature (15–30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.