Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brilliant Green, Certified, CI 42040, Ricca Chemical
SDP

Catalog No. RDCB053025F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCB053025F 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCB053025F Supplier Ricca Chemical Company Supplier No. RDCB053025F1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified laboratory-grade crystalline powder for research and industrial applications.

Chemical Identifiers

CAS 633-03-4
Molecular Formula C27H34N2O4S
Molecular Weight (g/mol) 482.639
InChI Key NNBFNNNWANBMTI-UHFFFAOYSA-M
PubChem CID 12449
IUPAC Name [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate
SMILES CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]

Specifications

CAS Min % 98.0
CAS Max % 100.0
Packaging Pouch
Color Yellow/Green
Quantity 25 g
Physical Form Crystalline Powder
Percent Purity ≥98%
Grade Laboratory
Chemical Name or Material Brilliant Green
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.