Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brilliant Sulfaflavine, MP Biomedicals™

Catalog No. ICN19507805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19507805 5 g
ICN19507825 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19507805 Supplier MP Biomedicals, Inc Supplier No. 0219507805
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2391-30-2
Molecular Formula C19H13N2NaO5S
Molecular Weight (g/mol) 404.372
InChI Key HYLDLLCHFLSKAG-UHFFFAOYSA-M
Synonym BSF
PubChem CID 23678860
ChEBI CHEBI:87226
IUPAC Name sodium;6-amino-2-(4-methylphenyl)-1,3-dioxobenzo[de]isoquinoline-5-sulfonate
SMILES CC1=CC=C(C=C1)N2C(=O)C3=CC=CC4=C3C(=CC(=C4N)S(=O)(=O)[O-])C2=O.[Na+]

Specifications

Formula Weight 418.4
Color Orange
Quantity 5 g
Physical Form Powder
Chemical Name or Material Brilliant Sulfaflavine
Recommended Storage Store at Room Temperature(15° to 30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.