Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brimonidine 98.0+%, TCI America™
SDP

Catalog No. B41321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41321G 1 g
B4132200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41321G Supplier TCI America Supplier No. B41321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59803-98-4
Molecular Formula C11H10BrN5
Molecular Weight (g/mol) 292.14
MDL Number MFCD00153878
InChI Key XYLJNLCSTIOKRM-UHFFFAOYSA-N
Synonym brimonidine, bromoxidine, mirvaso, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl-6-quinoxalinamine, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl quinoxalin-6-amine, 5-bromo-6-2-imidazolin-2-ylamino quinoxaline, unii-e6gnx3hhte, brimonidine inn:ban, 3h brimonidine, 5-bromo-6-imidazolin-2-ylamino quinoxaline
PubChem CID 2435
ChEBI CHEBI:3175
IUPAC Name 5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine
SMILES BrC1=C2N=CC=NC2=CC=C1NC1=NCCN1

Specifications

Melting Point 247°C
Quantity 1 g
UN Number 2811
Formula Weight 292.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Brimonidine
RTECSNumber VD1200000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.