Learn More
Apexbio Technology LLC Pemetrexed 150399-23-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A43905.1
Encompass_Preferred
Pemetrexed (CAS 150399-23-8) is a folate antagonist that targets multiple enzymes involved in the de novo synthesis of purine and pyrimidine nucleotides including thymidylate synthase (TS) dihydrofolate reductase (DHFR) glycinamide ribonucleotide formyltransferase (GARFT) and aminoimidazole carboxamide ribonucleotide formyltransferase (AICARFT) Structurally related to the antifolate compound lometrexol Pemetrexed disrupts nucleotide biosynthesis and subsequently inhibits DNA replication in proliferating cells It has demonstrated enzymatic inhibition across several pathways though exhibits limited cytotoxic activity in a range of tumor cell types such as non-small cell lung cancer mesothelioma breast colorectal cervical head and neck and bladder cancers This compound is widely used in preclinical studies of folate pathway inhibition and antimetabolite chemotherapy research
Catalog No. 50-000-96120
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.