Learn More
Britton-Robinson Buffer, pH 1.98, adjusted to 0.1 M Ionic Strength, Ricca Chemical

$209.00 - $338.00
Specifications
| Chemical Name or Material | Britton-Robinson Buffer |
|---|---|
| Color | Colorless Liquid |
| CAS | 7732-18-5 , 7447-40-7 , 7664-38-2 , 10043-35-3 , 64-19-7 , 1310-73-2 |
| CAS Max % | 98.55, 0.6, 0.36, 0.23, 0.22, 0.04 |
| Grade | Laboratory |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
R1154200500
![]() |
Ricca Chemical Company
R1154200500A |
500 mL |
Each for $209.00
|
|
|||||
R11542001A
![]() |
Ricca Chemical Company
R11542001A |
1 L |
Each for $338.00
|
|
|||||
Safety and Handling
ShelfLife : 24 months
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.