Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Britton-Robinson Buffer, pH 2.87, adjusted to 0.1 M Ionic Strength, Ricca Chemical
SDP

Catalog No. 11542916 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-542-916 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-542-916 Supplier Ricca Chemical Company Supplier No. 1154.2916
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

pH Reference Standard

Specifications

Color Colorless
pH 2.87
Physical Form Liquid
Quantity 500 mL
Packaging Natural Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.