Learn More
Applied Biosystems™ BRKT, System Power Supply
Description
This is a spare part for the 310 Genetic Analyzer.
Specifications
Specifications
| Quantity | 1 unit |
| For Use With (Equipment) | 310 Genetic Analyzer |
| Product Type | System Power Supply |
| Shipping Condition | Room Temperature |
| Content And Storage | Store at room temperature. |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.