Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BRL 50481
GREENER_CHOICE

Catalog No. 223710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-371-0 10 mg
22-375-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-371-0 Supplier Tocris Bioscience™ Supplier No. 2237/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective PDE7 inhibitor

Selective, substrate-competitive inhibitor of phosphodiesterase (PDE) 7 (Ki = 180 nM). Displays > 200-fold selectivity over PDE1B, PDE1C, PDE2, PDE3, PDE4A4 and PDE5.

Chemical Identifiers

CAS 433695-36-4
Molecular Formula C9H12N2O4S

Specifications

Quantity 10 mg
Formula Weight 244.26
Purity >99%
Molecular Weight (g/mol) 244.265
Chemical Name or Material BRL 50481
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.