Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bromacil Monoclonal Antibody (4G4)
GREENER_CHOICE

Catalog No. PIMA549732
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549732 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549732 Supplier Invitrogen™ Supplier No. MA549732
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Bromacil is a herbicide that is commonly used to control weeds in a variety of settings, including agricultural fields, roadsides, and industrial areas. It works by inhibiting photosynthesis in plants, which ultimately leads to their death. Bromacil is typically applied to the soil or sprayed directly onto the target weeds, and it is most effective when used in combination with other weed control methods, such as mowing or cultivation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bromacil
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 4G4
Concentration 1.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of BMC hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.