Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromate-Bromide Solution, 0.0200 N (N/50), Ricca Chemical
SDP

Catalog No. R116400020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R116400020F 20 L Cubitainer
R1164000500 500 mL Amber Glass Bottle
R11640001C 1 L Amber Glass Bottle
R11640004C 4 L Amber Glass Bottle
R116400010F 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. R116400020F Supplier Ricca Chemical Company Supplier No. R116400020F
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Bromate-Bromide Solution
Color Colorless
Name Note 0.0200 N (N/50)
CAS 1-2-7758
Assay Percent Range 0.02000 ± 0.00002 N at 20°C
Synonym dihydrogen oxide, dihydrogen monoxide
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Odor Odorless
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
Quantity 20 L
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.