Learn More
Bromate-Bromide Solution, 0.500 N (N/2), 0.250 M as Br2, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Chemical Name or Material | Bromate-Bromide Solution |
| Color | Colorless |
| CAS | 7732-18-5, 7758-01-2, 7758-01-2 |
| CAS Min % | 92.0, 4.7, 1.28 |
| CAS Max % | 96.0, 4.89, 1.33 |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| Molecular Weight (g/mol) | 119 g/mol |
| Grade | Laboratory |
| Packaging | Cubitainer |
| Quantity | 20 L |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.