Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromfenac sodium, Thermo Scientific™

Catalog No. AC468920010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468920010 1 g
AC468922500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC468920010 Supplier Thermo Scientific Chemicals Supplier No. 468920010
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 91714-93-1
Molecular Formula C15H11BrNNaO3
Molecular Weight (g/mol) 356.15
MDL Number MFCD03701673
InChI Key HZFGMQJYAFHESD-UHFFFAOYSA-M
IUPAC Name sodium 2-[2-amino-3-(4-bromobenzoyl)phenyl]acetate
SMILES [Na+].NC1=C(CC([O-])=O)C=CC=C1C(=O)C1=CC=C(Br)C=C1

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color Yellow to Orange
Quantity 1 g
Chemical Name or Material Bromfenac sodium
Hazard Category Environment
Recommended Storage Normal conditions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.