Learn More
Medchemexpress LLC Bromo-PEG3-C2-acid | 782475-35-8 | MFCD22574820 | ≥97.0% | 285.13 g/mol | C9H17BrO5 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW039168500MG
Encompass_Preferred
Bromo-PEG3-C2-acid is a PEG-based PROTAC linker containing a brominated PEG3 spacer and a C2 carboxylic acid functional group. It is designed for use in assembling bifunctional degraders and other conjugation chemistries where a reactive halide and a terminal acid facilitate linker installation and coupling.
  • Peg-based linker with a bromine functional handle.
  • Terminal carboxylic acid for standard conjugation chemistries.
  • Designed for use in PROTAC synthesis and linker assembly.
  • High purity (≥97.0%).
  • Available in multiple pack sizes for scale flexibility.
Catalog No. 50-003-00391