Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromoacetaldehyde dimethyl acetal, 97%, Thermo Scientific™

Catalog No. AC221875000
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC221875000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC221875000 Supplier Thermo Scientific Chemicals Supplier No. 221875000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7252-83-7
Molecular Formula C4H9BrO2
Molecular Weight (g/mol) 169.018
InChI Key FUSFWUFSEJXMRQ-UHFFFAOYSA-N
Synonym bromoacetaldehyde dimethyl acetal, ethane, 2-bromo-1,1-dimethoxy, 2,2-dimethoxyethyl bromide, dimethylbromoacetal, 1-bromo-2,2-dimethoxyethane, 2,2-dimethoxybromoethane, 2-bromoacetaldehyde dimethyl acetal, bromoacetoaldehyde dimethyl acetal, bromoacetaldehyde dimethylacetal, acetaldehyde, bromo-, dimethyl acetal
PubChem CID 81672
IUPAC Name 2-bromo-1,1-dimethoxyethane
SMILES COC(CBr)OC

Specifications

Density 1.4300g/mL
Boiling Point 148.0°C to 150.0°C
Flash Point 51°C
Refractive Index 1.4450 to 1.4470
Linear Formula BrCH2CH(OCH3)2
Quantity 500 mL
Specific Gravity 1.43
Solubility Information Solubility in water: insoluble.
Formula Weight 169.02
Chemical Name or Material Bromoacetaldehyde dimethyl acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.