Learn More
R&D Systems™ BromoCatch™ Expression Plasmid (N-term, pCMV, HygB)
SDP
Catalog No. RDEHBC05050 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
RDEHBC05050 50 μg
1 options
Catalog No. RDEHBC05050 Supplier R&D Systems™ Supplier No. RDEHBC05050
Add to Cart
Add to Cart
This plasmid enables expression of proteins fused to the BromoCatchTM tag, a covalent self-labeling tag derived from a BET bromodomain.
Specifications
Product Line R&D Systems
Quantity 50 μg