Learn More
R&D Systems™ BromoCatch™ GFP Expression Plasmid (N-term, pTK, HygB)
SDP
Catalog No. RD10008938 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
RD10008938 50 μg
1 options
Catalog No. RD10008938 Supplier R&D Systems™ Supplier No. RDELBC05GFP050
Customers require a valid business account associated to their profile to access pricing and ordering options.
This plasmid enables expression of proteins fused to the BromoCatchTM tag, a covalent self-labeling tag derived from a BET bromodomain.
Specifications
Product Line R&D Systems
Quantity 50 μg