Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromochloroacetic Acid Solution, 1000μg/mL in tert-butylmethyl ether, Ultra Scientific

Catalog No. USEPA1201
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA-1201 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-EPA-1201 Supplier Agilent Technologies Supplier No. EPA12011
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1634-04-4 , 5589-96-8
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

CAS Max % 99.86, 0.13
Analyte or Component Names tert-butylmethyl ether (MTBE),bromoacetic acid
Element Tert-butylmethyl ether, Bromoacetic Acid
Solvent or Matrix MTBE
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Bromochloroacetic Acid Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.