Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green, ACS Reagent (Sultone) not water soluble, Reagents

Catalog No. 01336780
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-780 5 g
01-336-782 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-780 Supplier Reagents Holdings Llc Supplier No. C61055005C2
Add to Cart
Add to Cart

Bromocresol Green, ACS Reagent (Sultone) not water soluble, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 76-60-8
Molecular Formula C21H14Br4O5S
Molecular Weight (g/mol) 698.01
InChI Key FRPHFZCDPYBUAU-UHFFFAOYSA-N
Synonym Bromocresol blue, Bromcresol Green, Bromcresol Blue
IUPAC Name 3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
SMILES CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C

Specifications

Grade ACS
Packaging Glass Bottle
Specific Gravity 0.79
Color Brown to Yellow
Quantity 5 g
Physical Form Solid
Chemical Name or Material Bromocresol Green, ACS Reagent (Sultone) not water soluble
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.