Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green-Methyl Red Mixed Indicator, Ricca Chemical
SDP

Catalog No. 12151 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
60 mL
120 mL
500 mL
4 L
Packaging:
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
12151 4 L Natural Poly Bottle
R121500060A 60 mL Natural Poly Bottle
1215-4 120 mL Natural Poly Bottle
1215-16 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12151 Supplier Ricca Chemical Company Supplier No. 12151
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Bromocresol Green-Methyl Red Mixed Indicator
CAS 7732-18-5, 62625-32-5, 845-10-3
CAS Min % 98.0, 0.1, 0.02
CAS Max % 100.0, 0.1, 0.02
Grade Indicator
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Dark Brown
Quantity 4 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.