Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green, Methyl Red TS, Reagents

Catalog No. 01335842
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-842 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-842 Supplier Reagents Holdings Llc Supplier No. CB003800120A
Add to Cart
Add to Cart

Bromocresol Green - Methyl Red TS, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 7732-18-5 , 7732-18-5 , 493-52-7
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Bromcresol Green-Methyl Red, MRBG
IUPAC Name ethanol
SMILES CCO

Specifications

Boiling Point 75°C
Packaging Poly Bottle
Specific Gravity 0.83
Color Red
Quantity 120 mL
Physical Form Liquid
Grade USP
Chemical Name or Material Bromocresol Green - Methyl Red TS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.