Learn More
Abcam Bromocresol green, pH indicator, 25G
Supplier: Abcam AB14632425G
MW 698.02 g/mol. Triiphenylmethane dye used as a pH indicator. Visual transition interval: pH 3.8 (yellow) – 5.4 (blue). Also used as a tracking dye for DNA agarose gel electrophoresis.
The product is subject to the following: Abcam Restricted Use Statement
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.