Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Bromocresol Purple, pure, indicator grade

Catalog No. AC151330250
Change view
Click to view available options
Quantity:
5 g
25 g
140 g
1 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151330250 25 g Glass bottle
AC151330050 5 g Glass bottle
AC151331400 140 g Glass bottle
AC151330010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC151330250 Supplier Thermo Scientific Chemicals Supplier No. 151330250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 115-40-2 | C21H16Br2O5S | 540.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-40-2
Molecular Formula C21H16Br2O5S
Molecular Weight (g/mol) 540.22
MDL Number MFCD00011681
InChI Key ABIUHPWEYMSGSR-UHFFFAOYSA-N
Synonym Bromcresol Purple, BCP, 5', 5''-Dibromo-o-cresolsulfonephthalein
PubChem CID 8273
ChEBI CHEBI:86154
SMILES CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1

Specifications

Absorbance (20 mg/L in methanol) Complies, (20 mg/L in methanol) Complies at 0nm
Melting Point 245.0°C
Solubility Information Solubility in water: practically insoluble. Other solubilities: 75g/L in ethanol (20°C)
Infrared Spectrum Authentic
IUPAC Name 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Formula Weight 540.21
Grade Pure
Packaging Glass bottle
Visual Transition Interval from pH 5.2 (yellow) to pH 6.8 (purple)
Color Pink or Salmon
Quantity 25 g
Physical Form Powder
Chemical Name or Material Bromocresol Purple
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.