Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocyclopropane 98.0+%, TCI America™
SDP

Catalog No. C106725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C106725G 25 g
C10675G 5 g
C1067250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C106725G Supplier TCI America Supplier No. C106725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4333-56-6
Molecular Formula C3H5Br
Molecular Weight (g/mol) 120.977
MDL Number MFCD00001271
InChI Key LKXYJYDRLBPHRS-UHFFFAOYSA-N
Synonym cyclopropyl bromide, cyclopropane, bromo, cyclopropylbromide, bromo-cyclopropan, bromo-cyclopropane, cyclopropylbrornide, bromanylcyclopropane, pubchem14417, bromocyclopropane, acmc-1aii5
PubChem CID 78037
IUPAC Name bromocyclopropane
SMILES C1CC1Br

Specifications

Color Yellow
Boiling Point 69°C
Quantity 25 g
UN Number 1993
Formula Weight 120.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromocyclopropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.