Learn More
R&D Systems™ Bromodeoxyuridine/BrdU MCF-7 Cell Line Slides (Adult Adenocarcinoma)- Paraffin
Shop All Novus Products

Description
- These BrdU-labeled MCF-7 cell line slides (Adult Adenocarcinoma) act as a reliable control for immunohistochemical and immunocytochemical staining procedures.
- Cell Line: MCF-7
- Species: Human
- Tissue: Breast
- Condition: Adenocarcinoma
- Life Stage: Adult
- Applications: Immunocytochemistry/Immunofluorescence, Immunohistochemistry
Specifications
Specifications
| Product Type | Cell Line Slide |
| Target Species | Human |
| Quantity | 5 Slides |
| For Use With (Application) | Immunohistochemistry (IHC), Immunocytochemistry/Immunofluorescence |
| Product Line | Novus |
| Species | Human |
| Target Specificity | Bromodeoxyuridine/BrdU |
| Content And Storage | Store at -20°C. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.