Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Bromodifluoromethyl)trimethylsilane 98.0+%, TCI America™
SDP

Catalog No. B43251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43251G 1 g
B43255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43251G Supplier TCI America Supplier No. B43251G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 115262-01-6
Molecular Formula C4H9BrF2Si
Molecular Weight (g/mol) 203.102
MDL Number MFCD18641931
InChI Key WDZVWBWAUSUTTO-UHFFFAOYSA-N
Synonym Bromodifluoro(trimethylsilyl)methane
PubChem CID 14639950
IUPAC Name [bromo(difluoro)methyl]-trimethylsilane
SMILES C[Si](C)(C)C(F)(F)Br

Specifications

Color Colorless
Boiling Point 108°C
Quantity 1 g
UN Number 1993
Formula Weight 203.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (Bromodifluoromethyl)trimethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.