Learn More
Selleck Chemical LLC Bromoenol lactone-E1878-1G
SDP
Supplier:  Selleck Chemical LLC E18781G
Encompass_Preferred
Bromoenol lactone((6E)-Bromoenol lactone) is a selective irreversible potent inhibitor of calcium-independent phospholipase A2 (iPLA2 ) with an IC50 value of 7 M It prevents antigen-stimulated exocytosis in mast cells without hindering Ca2 influx
Catalog No. 50-313-7710
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.