Learn More
Bromoethane, 98%
Description
Bromoethane is used in organic synthesis for the introduction of ethyl group. It is widely used as solvent, as an anesthetic in medicine and as a refrigerant. It is an important component of antiknock fluid in gasoline. It is also also used in the manufacture of pharmaceuticals like vitamin A as well as in the manufacture of flame retardants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bromoethane is used in organic synthesis for the introduction of ethyl group. It is widely used as solvent, as an anesthetic in medicine and as a refrigerant. It is an important component of antiknock fluid in gasoline. It is also also used in the manufacture of pharmaceuticals like vitamin A as well as in the manufacture of flame retardants.
Solubility
Miscible with ethanol, ether, chloroform and organic solvents.
Notes
Light sensitive. Store in cool place. Incompatible with strong bases, strong oxidizing agents, magnesium, potassium, calcium, sodium oxides, aluminum, zinc and alkali metals.
Specifications
Specifications
| Melting Point | -119°C |
| Density | 1.461 |
| Boiling Point | 37°C to 39°C |
| Flash Point | −23°C (−9°F) |
| Odor | Ether-like |
| Assay Percent Range | ≥98% |
| Refractive Index | 1.4235 |
| Linear Formula | BrCH2CH3 |
| Quantity | 2500 g |
| UN Number | UN1891 |
| Show More |
Safety and Handling
| Hazard Category | H225-H302+H332-H351 |
| Hazard Statement | GHS H Statement H225-H351-H302-H332 Highly flammable liquid and vapour. Suspected of causing cancer. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P201-P202-P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P308+P313-P330-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ETHYL BROMIDE |
| EINECSNumber | 200-825-8 |
| RTECSNumber | KH6475000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.