Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromoform, 96%, stabilized

Catalog No. AC158200010
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
5 L
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158200010 1 L Glass bottle
AC158202500 250 mL Glass bottle
AC158200025 2.5 L Plastic drum
AC158200050 5 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC158200010 Supplier Thermo Scientific Chemicals Supplier No. 158200010
Add to Cart

CAS: 75-25-2 | CHBr3 | 252.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-25-2 , 64-17-5
Molecular Formula CHBr3
Molecular Weight (g/mol) 252.73
MDL Number MFCD00000128
InChI Key DIKBFYAXUHHXCS-UHFFFAOYSA-N
Synonym tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
PubChem CID 5558
ChEBI CHEBI:38682
IUPAC Name bromoform
SMILES C(Br)(Br)Br

Specifications

Melting Point 8.0°C
Color Colorless to Orange-Yellow
Density 2.6300g/mL
Boiling Point 150.0°C to 151.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CHBr3
Quantity 1 L
Beilstein 01,68
Fieser 12,74
Specific Gravity 2.63
Merck Index 15,143
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,benzene,chloroform,ether,,petroleum ether,acetone and oils
Stabilizer 3 to 4 wt% ethanol
Formula Weight 252.73
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Bromoform
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-854-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.