Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromoform, 99+%, stabilized

Catalog No. AC220691000
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC220691000 100 mL Glass bottle
AC220690250 25 mL Glass bottle
AC220695000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC220691000 Supplier Thermo Scientific Chemicals Supplier No. 220691000
Add to Cart

CAS: 75-25-2 | CHBr3 | 252.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-25-2 , 64-17-5
Molecular Formula CHBr3
Molecular Weight (g/mol) 252.73
InChI Key DIKBFYAXUHHXCS-UHFFFAOYSA-N
Synonym tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
PubChem CID 5558
ChEBI CHEBI:38682
IUPAC Name bromoform
SMILES C(Br)(Br)Br

Specifications

Melting Point 8°C
Color White
Density 2.8900g/mL
Boiling Point 150°C to 151°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (excluding stabilizer) (GC)
Packaging Glass bottle
Linear Formula CHBr3
Quantity 100 mL
Beilstein 01, 68
Fieser 12,74
Specific Gravity 2.89
Merck Index 15, 1430
Solubility Information Solubility in water: slightly soluble
Stabilizer 1.5 wt% max. ethanol
Formula Weight 252.73
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Bromoform
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with wa
EINECSNumber 200-854-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.