Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromohydroquinone 90.0+%, TCI America™
SDP

Catalog No. B160025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B160025G 25 g
B16005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B160025G Supplier TCI America Supplier No. B160025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 583-69-7
Molecular Formula C6H5BrO2
Molecular Weight (g/mol) 189.008
MDL Number MFCD00041747
InChI Key REFDOIWRJDGBHY-UHFFFAOYSA-N
Synonym bromohydroquinone, 2-bromohydroquinone, 1,4-benzenediol, 2-bromo, 2-bromoquinol, 2-bromo-1,4-benzenediol, hydroquinone, bromo, bromoquinol, 4-hydroxy bromophenol, 2-bromo-benzene-1,4-diol
PubChem CID 68502
IUPAC Name 2-bromobenzene-1,4-diol
SMILES C1=CC(=C(C=C1O)Br)O

Specifications

Quantity 25 g
Formula Weight 189.01
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bromohydroquinone
RTECSNumber CZ8920000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.