Learn More
Sigma Organic Chemistry Bromomethyl cyclopentan | 100mg | MFCD09263483
Supplier: Sigma Organic Chemistry CDS005521100MG

Bromomethyl cyclopentan , 100mg
About this Item:
MDL #: MFCD09263483
Molecular Weight: 163.06
UNSPSC Code: 12352200
Chemical Formula: C6H11Br
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.