Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromopentamethylbenzene 97.0+%, TCI America™
SDP

Catalog No. B415425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B415425G 25 g
B41545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B415425G Supplier TCI America Supplier No. B415425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5153-40-2
Molecular Formula C11H15Br
Molecular Weight (g/mol) 227.15
MDL Number MFCD00013523
InChI Key XPDQRULPGCFCLX-UHFFFAOYSA-N
Synonym bromopentamethylbenzene, benzene, bromopentamethyl, pentamethylbromobenzene, 1-bromopentamethylbenzene, 1-bromo-2,3,4,5,6-pentamethyl-benzene, brompentamethylbenzol, maybridge1_002545, ksc273s6f, acmc-1b227, 5-bromo-1,2,3,4,6-pentamethylbenzene
PubChem CID 78831
IUPAC Name 1-bromo-2,3,4,5,6-pentamethylbenzene
SMILES CC1=C(C)C(C)=C(Br)C(C)=C1C

Specifications

Melting Point 162°C
Color White-Yellow
Boiling Point 290°C
Quantity 25 g
Formula Weight 227.15
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bromopentamethylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.