Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromophenol Blue (Certified ACS), Fisher Chemical

Catalog No. B3925 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B392-5 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B392-5

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 115-39-9
Molecular Formula C19H10Br4O5S
Molecular Weight (g/mol) 669.96
MDL Number MFCD00005875
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
Synonym 4,4'-(3H-2,1-Benzoxathiol-3-ylidene) bis-[2,6-dibromophenol] S,S-Dioxide, 4, 4'-(3H-2, 1-Benzoxathiol-3-ylidene) bis-[2
PubChem CID 8272
ChEBI CHEBI:59424
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Melting Point 279°C
Clarity Pass Test
Linear Formula C6H4SO2OC(C6H2-3, 5-Br2-4-OH)2
Formula Weight 669.96
Grade Certified ACS
Packaging Glass Bottle
Vapor Pressure Vapor Pressure: Negligible
Visual Transition Interval Pass Test
Color Violet
Quantity 5 g
Physical Form Solid
Chemical Name or Material Bromophenol Blue, pH Indicator
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: CAUTION!
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated.
NFPA
Health:0 Flammability:0 Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.