CAS RN 115-39-9
Thermo Scientific Chemicals Bromophenol Blue
CAS: 115-39-9 Molecular Formula: C19H10Br4O5S Molecular Weight (g/mol): 669.96 MDL Number: MFCD00005875 InChI Key: UDSAIICHUKSCKT-UHFFFAOYSA-N Synonym: Bromphenol Blue PubChem CID: 8272 ChEBI: CHEBI:59424 IUPAC Name: 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol SMILES: C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
Thermo Scientific Chemicals Bromophenol Blue, ACS
CAS: 115-39-9 Molecular Formula: C19H10Br4O5S Molecular Weight (g/mol): 669.96 MDL Number: MFCD00005875 InChI Key: UDSAIICHUKSCKT-UHFFFAOYSA-N PubChem CID: 8272 ChEBI: CHEBI:59424 IUPAC Name: 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,lambda {6}-benzoxathiol-3-yl]phenol SMILES: C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
Thermo Scientific Chemicals Bromophenol Blue, ACS reagent
CAS: 115-39-9 Molecular Formula: C19H10Br4O5S Molecular Weight (g/mol): 669.96 MDL Number: MFCD00005875 InChI Key: UDSAIICHUKSCKT-UHFFFAOYSA-N Synonym: 3',3'',5',5''-Tetrabromophenolsulfonephthalein,BPB,3', 3'', 5' PubChem CID: 8272 ChEBI: CHEBI:59424 IUPAC Name: 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1,lambda{6}-benzoxathiol-3-yl]phenol SMILES: C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
Thermo Scientific Chemicals Bromophenol Blue, pure, indicator
CAS: 115-39-9 Molecular Formula: C19H10Br4O5S Molecular Weight (g/mol): 669.96 MDL Number: MFCD00005875 InChI Key: UDSAIICHUKSCKT-UHFFFAOYSA-N Synonym: 3',3'',5',5''-Tetrabromophenolsulfonephthalein,BPB,3', 3'', 5' PubChem CID: 8272 ChEBI: CHEBI:59424 IUPAC Name: 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol SMILES: C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
Bromophenol Blue, Sodium Salt, MP Biomedicals™
ChEBI: CHEBI:59424
Bromophenol Blue, pure, 0.1% in water
CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol
Bromophenol Blue, Sulfone Form, MP Biomedicals
CAS: 115-39-9 Molecular Formula: C19H10Br4O5S Molecular Weight (g/mol): 669.96 InChI Key: UDSAIICHUKSCKT-UHFFFAOYSA-N Synonym: 3,3′,5,5′-Tetrabromophenolsulfonphthalein PubChem CID: 8272 ChEBI: CHEBI:59424 IUPAC Name: 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol SMILES: C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
Bromophenol Blue (0.1% in ca. 50% Ethanol) [for Titration], TCI America™
CAS: 115-39-9 Molecular Formula: C19H10Br4O5S Molecular Weight (g/mol): 669.96 MDL Number: MFCD00005875 InChI Key: UDSAIICHUKSCKT-UHFFFAOYSA-N Synonym: bromophenol blue,bromphenol blue,albutest,tetrabromphenol blue,3,3',5,5'-tetrabromophenolsulfonphthalein,3',3,5',5-tetrabromophenolsulfonephthalein,ccris 5487,bromophenol blue bpb,3',3,5',5-tetrabromophenolsulfophthalein,bromphenol blue sultone form PubChem CID: 8272 ChEBI: CHEBI:59424 IUPAC Name: 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol SMILES: C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
Bromophenol Blue, TCI America™
CAS: 115-39-9 Molecular Formula: C19H10Br4O5S Molecular Weight (g/mol): 669.96 MDL Number: MFCD00005875 InChI Key: UDSAIICHUKSCKT-UHFFFAOYSA-N PubChem CID: 8272 ChEBI: CHEBI:59424 IUPAC Name: 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol SMILES: C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
Bromophenol Blue, ACS Reagent (Sultone) not water soluble, Reagents
Bromophenol Blue, ACS Reagent (Sultone) not water soluble, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Bromophenol Blue, Indicator, Reag. Ph. Eur., Solstice
CAS: 115-39-9 Molecular Formula: C19H10Br4O5S Molecular Weight (g/mol): 669.96 MDL Number: MFCD00005875 InChI Key: UDSAIICHUKSCKT-UHFFFAOYSA-N PubChem CID: 8272 ChEBI: CHEBI:59424 IUPAC Name: 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol SMILES: C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
Bromophenol Blue Indicator Solution, Lovibond™
Bromophenol blue indicator solution for water analysis applications.
- 1
- 2