Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromophenol Blue, Sodium Salt, MP Biomedicals™

Catalog No. 0215250625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
50 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-152-50625 25 g
ICN15250605 5 g
04-805-732 10 g
02-152-50650 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-152-50625 Supplier MP Biomedicals, Inc Supplier No. 0215250625
Add to Cart
Add to Cart

Bromophenol Blue, Sodium Salt is a tracking dye.

  • Bromophenol blue migrates at approximately the same rate as 300-500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels.
  • It is a tracking dye for alkaline and neutral buffer systems, as well as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 34725-61-6
Molecular Formula C19H9Br4NaO5S
Solubility Information Soluble in water (30 mg/mL-clear, blueish violet solution) or ethanol (7 mg/mL).
InChI Key KGANHHDZDYZJEM-UHFFFAOYNA-M
SMILES [Na+].OC1=CC(Br)=C(C(Br)=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=C(Br)C=C([O-])C=C1Br
IUPAC Name sodium 3,5-dibromo-4-[3-(2,6-dibromo-4-hydroxyphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]benzen-1-olate
Molecular Weight (g/mol) 691.94
Grade ACS
Quantity 25 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.