Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromophenol Red 80.0+%, TCI America™
SDP

Catalog No. B06321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B06321G 1 g
B063225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B06321G Supplier TCI America Supplier No. B06321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2800-80-8
Molecular Formula C19H12Br2O5S
Molecular Weight (g/mol) 512.168
MDL Number MFCD00023014
InChI Key OYCLSQDXZMROJK-UHFFFAOYSA-N
PubChem CID 76047
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C=C3)O)Br)C4=CC(=C(C=C4)O)Br

Specifications

Formula Weight 512.17
Percent Purity ≥80.0% (HPLC)
Quantity 1 g
Physical Form Crystal/Powder
Chemical Name or Material Bromophenol Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.