Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Bromothymol Blue, pH Indicator, 624.38 g/mol, 5 g

Catalog No. 502704753
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4753 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4753 Supplier Abcam Supplier No. AB1462935G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

pH indicator for weak acids and bases with a range of 6.0 to 7.6, also used as a microscopic stain.

  • Molecular Weight: 624.38 g/mol
  • pH Indicator Range: 6.0 (yellow) to 7.6 (blue)
  • Applications: pH indicator for weak acids and bases; stain for defining cell walls or nuclei under the microscope
  • Composition: 100% Bromothymol blue free acid

Specifications

Shipping Condition Ambient
Detection Method Colorimetric
Buffer 100% Bromothymol blue free acid
Content And Storage Store at ambient temperature.
Detection Location In-Cell Detection
Form Powder
Formulation 100% Bromothymol blue free acid
Quantity 5 g
For Use With (Application) pH indication, Microscopy-staining
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.