Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Bromothymol Blue Solution, 0.04%
SCI_ED

Catalog No. S25207C Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 mL
30 mL
100 mL
500 mL
1 L
Packaging:
0.04%
Lab Grade - Solution
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25207C 1 L 0.04%
S05149 30 mL Lab Grade - Solution
S25207A 100 mL 0.04%
S25207B 500 mL 0.04%
S25207 10 mL 0.04%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S25207C

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

CO2/pH indicator

This product is for educational use - no C of A is available.

Specifications

Chemical Name or Material Bromothymol Blue Solution, 0.04%
CAS 62625-28-9, 7732-18-5
Assay Percent Range 0.04% w/v
Formula Weight 646.37
Packaging 0.04%
Density 1g/mL
pH Color Changes pH 6.0-7.6 (yellow-blue)
Color Green
Quantity 1 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.