Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromothymol Blue Sultone Form, ACS Reagent, Reag. Ph. Eur., Solstice

Catalog No. 60046154 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
1 kg
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-154 1 kg Glass Bottle
60-046-156 5 g Glass Bottle
60-046-155 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-046-154 Supplier Solstice Supplier No. 327141KG
Add to Cart
Add to Cart

Indicator, ACS Reagent, Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-59-5
Molecular Formula C27H28Br2O5S
Molecular Weight (g/mol) 624.384
MDL Number MFCD00005872
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
Synonym 3′,3″-Dibromothymolsulfonphthalein
PubChem CID 6450
ChEBI CHEBI:86155
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br

Specifications

UN Number NONH for all modes of transport
Beilstein 373934
Formula Weight 624.38g/mol
Grade ACS Reagent
Packaging Glass Bottle
Suitability Complies for clarity of solution
Visual Transition Interval 5.8 to 7.4
Color Violet
Quantity 1 kg
Physical Form Powder
Chemical Name or Material Bromothymol Blue Sultone Form, Reag. Ph. Eur.
Show More Show Less
EINECSNumber 200-971-2
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.