Learn More
StatLab BROWN & BRENN STAIN KIT
SDP
Supplier:  StatLab KTBBR
Encompass_Preferred
This kit stains for gram positive and gram negative bacteria in just 10 minutes. Stain kit includes Gentian Violet 100ml, 1% Safranin O, Universal Iodine Solution, ADVANCED Picric Acid/Acetone, and Gram's Decolorizer.
Catalog No. NC9765115
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.