Learn More
Perkin Elmer US LLC Brownlee Spheri-5 Cyano Cartridge Column - 220 mm x 4.6 mm I.D.
Supplier:  Perkin Elmer US LLC 07110047
Encompass_Preferred
Recommended for the normal-phase separation of smaller, relatively polar compounds. Offers alternative selectivity in reversed phase to alkyl phases.
Catalog No. 50-211-3555
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.