Learn More
Perkin Elmer US LLC Brownlee Spheri-5 RP-8 Cartridge Column - 30 mm x 4.6 mm I.D., Pkg. 2
Supplier:  Perkin Elmer US LLC 07110001
Encompass_Preferred
Monofunctional bonded C8 phase for general purpose reversed phase small molecule applications.
Catalog No. 50-211-3506
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.