Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRPF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527783 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527783 Supplier Invitrogen™ Supplier No. PA527783
  • $620.00
    $489.65 / Each of 1

    Save $130.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, BRPF1 tansfected 293T cell. Predicted reactivity: Mouse (97%), Rat (96%), Chicken (89%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

BRPF1 (peregrin) is a component of the MOZ/MORF complex. This complex has histone H3 acetyltransferase activity. BRPF1 preferentially mediates histone H3-K23 acetylation and positively regulates the transcription of RUNX1 and RUNX2. Mutations affecting the gene can result in intellectual developmental disorder with dysmorphic facies and ptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRPF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene BRPF1
Gene Accession No. B2RRD7, P55201
Gene Alias 4833438B11Rik; 4930540D11Rik; BR140; bromodomain and PHD finger containing 1; bromodomain and PHD finger containing, 1; bromodomain and PHD finger-containing protein 1; bromodomain-containing protein, 140kD; Brpf1; Brpf2; Peregrin; Protein Br140
Gene Symbols BRPF1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human Peregrin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7862, 78783
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.