Learn More
TARGETMOL CHEMICALS INC BRUCEINE D 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TWS2045
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Bruceine D from Brucea javanica may have the potential to be used as a natural viricide or a lead compound for new viricides. 2. Bruceine D inhibits the growth of three pancreatic cancer cell lines i.e. PANC-1 SW199 and CAPAN-1; induces cytotoxicity in Capan-2 cells via the induction of cellular apoptosis involving the mitochondrial pathway. purity: 99%
Catalog No. 50-257-6145
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.