Learn More
Liofilchem™ Brucella Blood Agar With Hemin and Vitamin K1
SDP
Catalog No. 21403468 Shop All Liofilchem Products
Change view
Click to view available options
Quantity:
20 Plates
Catalog No. Quantity
21-403-468 20 Plates
1 options
Catalog No. 21-403-468 Supplier Liofilchem™ Supplier No. 10245
Add to Cart
Add to Cart
A medium for isolation of obligate anaerobes and MIC determination with Hemin and Vitamin K1
  • Medium for the isolation and subculture of anaerobes from clinical specimens
  • This medium is intended as an aid in the diagnosis, requiring further tests to complete the diagnostic results
Specifications
Manufacturing Origin Italy
Product Type Prepared Microbiology Plate
Sterility Non-sterile
Target Organism Class Anaerobes
Test Type Microbial Cultivation
Quantity 20 Plates
Diameter (Metric) 90 mm