Learn More
Bioworld Brucella Broth, 2 kg
Supplier:  Bioworld 306200163
Encompass_Preferred
Brucella brothRecommended for the cultivation of Brucella species and other fastidious microoorganismspH: 7.0Dehydrated Appearance: light to medium beige, homogeneous powderPrepared Appearance: gold to beige brothSoluble in waterContains:Pancreatic Digest of Casein Peptic Digest of Animal Tissue Yeast Extract Sodium Chloride Dextrose Sodium Bisulfite
Catalog No. 50-254-290
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.