Learn More
ATCC Brucella canis Carmichael and Bruner (ATCC 23365) - Frozen research culture, BSL 3
Supplier: ATCC 23365
Brucella canis is supplied as a frozen, research-only bacterial culture intended for laboratory study of zoonotic infection. Due to its Biosafety Level 3 classification, this strain must be handled in appropriate containment; growth conditions and detailed handling instructions are provided to qualified purchasers.
- Type strain of Brucella canis.
- Isolation source: allantoic fluid of an aborted beagle puppy.
- Format: frozen culture for research use.
- Storage: store at -80°C or colder.
- Biosafety: handled at Biosafety Level 3 facilities.
- Intended use: laboratory research on zoonotic disease.
- Growth conditions: details provided to qualified purchasers.
- Viability warranty: warranted for 30 days from shipment when stored per instructions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.